Learn More
Abnova™ Global Di-Methyl Histone H3-K27 Quantification Kit (Colorimetric)
Use for measuring global Di-Methylation of Histone H3-K27
Manufacturer:  Abnova™ KA0602
Catalog No. 89-027-163
Add to cart
Specifications
Specifications
Global Di-Methyl Histone H3-K27 Quantification Kit (Colorimetric) is use for measuring global di-methylation of histone H3-K27.
Quantification
Global Di-Methyl Histone H3-K27 Quantification Kit (Colorimetric)
1 Kit
Colorimetric
20mL C1 (Wash Buffer 10X), 12mL C2 (Antibody Buffer), 10μL C3 (detection Antibody, 1mg/mL), 10μL C4 (color developer), 6mL C5 (stop solution), 20μL standard control (100μg/mL), 98-wel
Global Di-Methyl Histone H3-K27 quantification kit
Store C3 and Standard control at -20°C. Store all other components at 4°C away from light.