Learn More
Abnova™ Global Histone H3 Phosphorylation (Ser28) Assay Kit (Colorimetric)
For measuring global Phospho Histone H3 (Ser28)
Manufacturer:  Abnova™ KA0705
Catalog No. 89-027-062
Add to cart
Specifications
Specifications
Global Histone H3 Phosphorylation (Ser28) Assay Kit (Colorimetric) is use for measuring global phospho histone H3 (Ser28).
Quantification
Global Histone H3 Phosphorylation (Ser28) Assay Kit (Colorimetric)
1 Kit
Colorimetric
20mL C1 (Wash Buffer 10X), 12mL C2 (Antibody Buffer), 10μL C3 (detection Antibody, 1mg/mL), 10mL C4 (color developer), 6mL C5 (stop solution), 20μL standard control (100μg/mL), 98-well sample strips (w
Global Histone H3 Phosphorylation (Ser28) assay kit
Store C3 and Standard control at -20°C. Store all other components at 4°C away from light.