Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Global Mono-Methyl Histone H4K20 Quantification Kit (Colorimetric)
To measure global mono-methylation of histone H4K20
Supplier: Abnova™ KA1517
Specifications
20 to 2000ng/well | |
Global Mono-Methyl Histone H4K20 Quantification Kit (Colorimetric) is used for measuring global mono-methylation of histone H4K20. | |
Quantification | |
1 Kit | |
50 μL |
Store the standard at -20°C. Store all other components at 4°C away from light. | |
Colorimetric | |
10mL C1 (wash buffer 10X), 6mL C2 (antibody buffer), 5μL C3 (detection antibody, 1mg/mL), 5mL C4 (color developer), 3mL C5C5 (stop solution), 10μL standard control (100μg/mL), 5μL sig | |
Histone Extract | |
Quantification Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction