Learn More
Abnova™ Global Tri-Methyl Histone H4K20 Quantification Kit (Colorimetric)
To measure global tri-methylation of histone H4K20
Manufacturer:  Abnova™ KA1521
Catalog No. 89-028-216
Add to cart
Specifications
Specifications
20 to 2000ng/well
Quantification
1 Kit
50μL
Colorimetric
Global Tri-Methyl Histone H4K20 Quantification Kit (Colorimetric) is used for measuring global tri-methylation of histone H4K20.
10mL C1 (wash buffer 10X), 6mL C2 (antibody buffer), 5μL C3 (detection antibody, 1mg/mL), 5mL C4 (color developer), 3mL C5 (stop solution), 10μL standard control (100μg/mL), 5μL signa
Histone extract
Store the standard at -20°C. Store all other components at 4°C away from light.
Global Tri-methyl Histone H4K20 Quantification Kit