Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Glucose-6-Phosphate Dehydrogenase Assay Kit

Simple, sensitive and rapid assay detects the activity of G6PDH in a variety of samples.

Supplier:  Abnova™ KA0880

Catalog No. 89-024-435


Due to product restrictions, please to purchase or view availability for this product.


Includes:
  • G6PDH Assay buffer 25mL
  • G6PDH Substrate (lyophilized), 1 vial
  • G6PDH Developer (lyophilized), 1 vial
  • G6PDH Positive control (lyophilized), 1 vial
  • NADH Standard (0.5μmol) (lyophilized), 1 vial

Description

Description

In the assay, glucose-6-phosphate is oxidized with the generation of a product which is utilized to convert a nearly colorless probe to an intensely colored product with an absorbance at 450nm. The G6PDH Assay Kit can detect as low as 0.04mU G6PDH per well.
Specifications

Specifications

1 Kit
Store the kit at -20°C
Functional Study
Glucose-6-Phosphate Dehydrogenase Assay Kit is used for detecting G6PDH activity.
Colorimetric
0.04 mU
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit