Learn More
Abnova™ Glucose Assay Kit II
Provides direct measurement of glucose in various biological samples (e.g., serum, plasma, other body fluids, food, growth media, etc.). The method is rapid, simple, sensitive, and suitable for high throughput.
Manufacturer:  Abnova™ KA0832
Catalog No. 89-028-068
Add to cart
Description
Description
  • In this assay, glucose is specifically oxidized to generate a product which reacts with a dye to generate color (λ = 450nm) whose intensity is proportional to glucose concentration
  • Particularly suitable for serum and urine samples since it is unaffected by reducing substances which can interfere with other suppliers offering oxidase‐based kits
  • Also suitable for monitoring glucose level during fermentation and glucose feeding in protein expression processes
Specifications
Specifications
20 to 10000μM
Functional Study
1 Kit
Store the kit at -20°C.
Glucose Assay Kit II
Glucose Assay Kit II is used for measuring glucose.
25mL Glucose assay buffer, 1 vial Glucose substrate mix (lyophilized), 1 vial Glucose enzyme mix (lyophilized), 100μL Glucose standard (100mM)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric