Learn More
Abnova™ Glutathione Reductase Assay Kit
Glutathione Reductase Assay Kit is used for measuring GR activity.
Manufacturer:  Abnova™ KA0881
Catalog No. 89-024-436
Add to cart
Description
Description
  • A highly sensitive, simple, direct, and HTS-ready colorimetric assay for measuring GR activity in biological samples
  • Assay can detect 0.1 to 40 mU/ml GR in various samples
  • Kit has a detection sensitivity of approximately 0.5 mU/ml of GPx in samples
  • Store kit at -20°C; protect from light
Specifications
Specifications
1 Kit
Store the kit at -20°C.
0.1 to 40mU/mL
Glutathione Reductase Assay Kit is used for measuring GR activity.
Colorimetric
Functional Study