Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ GST Activity Assay Kit (Colorimetric)
To detect GST activity in crude cell lysate or purified protein fraction and also quantitate GST-tagged fusion protein, functional study
Supplier: Abnova™ KA0799
Specifications
Store the kit at -20°C. | |
Colorimetric | |
10mL GST assay buffer, 25mL GST sample buffer, 1mL GST substrate (CDNB), 34mg Glutathione (GSH) (lyophilized), 20μL GST positive control (0.625μg/μL) | |
Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |
GST Activity Assay Kit (Colorimetric) is used for detecting GST activity in crude cell lysate or purified protein fraction, and also quantitate GST-tagged fusion protein. | |
Functional Study | |
1 Kit | |
Activity Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction