Learn More
Abnova™ GST Activity Assay Kit (Fluorometric)
To detect the GST activity, functional study
Manufacturer:  Abnova™ KA0796
 View more versions of this product
Catalog No. 89-028-099
Add to cart
Specifications
Specifications
0.5 mU
Functional Study
1 Kit
Store the kit at -20°C.
GST Activity Assay Kit
GST Activity Assay Kit (Fluorometric) is used for detecting the GST activity.
10mL GST assay buffer, 25mL GST sample buffer, 200μL MCB substrate (in DMSO), 2 vials Glutathione (lyophilized), 50μL GST standard (5mU/μL)
Cell sample, Erythrocute sample, Plasma sample, Tissue sample
Fluorometric