Learn More
Abnova™ HAT Activity Assay Kit
To detect histone acetyltransferase (HAT) activity in mammalian samples, functional study
Manufacturer:  Abnova™ KA0782
Catalog No. 89-028-111
Add to cart
Specifications
Specifications
HAT Activity Assay Kit is used for detecting for HAT activity in mammalian samples.
7.5mL 2X HAT assay buffer, 1 vial HAT substrate I, 1 vial HAT substrate II, 1 vial NADH generating enzyme, 50μL NE (Nuclear Extract 4mg/mL), 1.8mL HAT Reconstitution buffer
Cell lysate, tissue lysate
Colorimetric
Functional Study
1 Kit
Store the kit at -80°C.
HAT Activity Assay Kit