Learn More
Abnova™ HAT Activity/Inhibition Assay Kit (Colorimetric)
Use for measuring HAT activity/inhibition
Manufacturer:  Abnova™ KA0626
Catalog No. 89-027-187
Add to cart
Specifications
Specifications
HAT Activity/Inhibition Assay Kit (Colorimetric) is use for measuring HAT activity/inhibition.
Functional Study
HAT Activity/Inhibition Assay Kit (Colorimetric)
1 Kit
Colorimetric
28mL HT1 (Wash Buffer 10X), 100μL HT2 (HAT Substrate, 20μg/mL), 50μL HT3 (HAT standard, 20μg/mL), 20μL HT4 (Acetyl Co-A, 30mM), 3mL HT5 (HAT Assay Buffer), 50
HAT activity/inhibition assay kit
Store HT2, HT3, HT4 and HT7 at -20°C away from light. Store HT6, HT8 and 8 well strips at 4°C away from light. Store all other components at room temperature.