Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ HDAC Activity Assay Kit (Colorimetric)
To analyze histone deacetylases (HDAC) activity, functional study
Supplier: Abnova™ KA0788
Specifications
Store the kit at -80°C. | |
Colorimetric | |
500μL HDAC substrate (Boc-Lys(Ac)-pNA, 10mM), 1mL HDAC assay buffer 10X, 1mL Lysine developer, 10μL HDAC inhibitor (Trichostatin A, 1mM), 50μL HeLa nuclear extract (5mg/mL), 20μL Deac | |
Biological Fluid, Cell Lysate, Plasma, Serum, Tissue Lysate, Urine |
HDAC Activity Assay Kit (Colorimetric) is used for analyzing HDAC activity. | |
Functional Study | |
1 Kit | |
Activity Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction