Learn More
Abnova™ HDAC Activity/Inhibition Assay Kit (Colorimetric)
Use for measuring HDAC activity/inhibition
Manufacturer:  Abnova™ KA0628
Catalog No. 89-027-189
Add to cart
Specifications
Specifications
HDAC Activity/Inhibition Assay Kit (Colorimetric) is use for measuring HDAC activity/inhibition.
Functional Study
HDAC Activity/Inhibition Assay Kit (Colorimetric)
1 Kit
Colorimetric
28mL H1 (Wash Buffer 10X), 3mL H2 (HDAC Assay Buffer), 100μL H3 (Biotinylated HDAC Substrate), 100μL H4 (HDAC inhibitor, 0.5mM), 50μL H5 (HDAC standard 20μg/mL), 50μ
HDAC activity/inhibition assay kit
Store H3, H4, H5 and H7 at -20°C away from light. Store H6, H8 and 8 well strips at 4°C away from light. Store all other components at room temperature.