Learn More
Abnova™ HDAC Inhibitor Drug Screening Kit
To screen histone deacetylases (HDAC) inhibitors, functional study
Manufacturer:  Abnova™ KA0789
Catalog No. 89-028-104
Add to cart
Specifications
Specifications
HDAC Inhibitor Assay Kit is used for screening HDAC inhibitors.
500μL HDAC substrate (Boc-Lys(Ac)-pNA, 10mM), 1mL HDAC assay buffer 10X, 1mL Lysine developer, 10μL HDAC inhibitor (Trichostatin A, 1mM), 10μL HeLa nuclear extract (5mg/mL), 20μL Deac
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
HDAC Inhibitor Assay Kit