Learn More
Abnova™ Hemin Assay Kit
Used for quantitating hemin and functional study
Manufacturer:  Abnova™ KA0871
Catalog No. 89-024-426
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
5 to 160pg
Functional Study
1 Kit
Store the kit at 4°C, protect from light. Probe to room temperature before use. Enzyme Mix dissolve in Hemin Assay Buffer, mix well. Store at -20°C.
Hemin Assay Kit
Hemin Assay Kit is used for quantitating hemin.
25mL Hemin assay buffer, 0.2mL Hemin probe (in DMSO), 1 vial enzyme mix, 1mL Hemin substrate, 1 vial Hemin standard (1nM, lyophilized)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine, food sample, growth medium
Colorimetric