Learn More
Abnova™ Hepatitis E virus IgM (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of HEV specific IgM in Human Serum and Plasma
Manufacturer:  Abnova™ KA1009
Catalog No. 89-028-181
Add to cart
Specifications
Specifications
HRP
HEV IgM (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of HEV specific IgM in human serum and plasma.
Qualitative
Hepatitis E virus IgM (Human) ELISA Kit
1 Kit
Store the kit at 4°C.
Human
Colorimetric
96-well Synthetic HEV polypeptide coated microwell, 12mL Conjugate, 1mL non reactive control, 1mL reactive control, 60mL sample diluent, 60mL Wash Buffer (20X), 10mL Substrate A, 10mL substrate B, 14mL stop solution
Hepatitis E Virus IgM (Human) ELISA kit
Plasma, Serum
100μL