Learn More
Abnova™ Histone Demethylase (H3-K9 Specific) Activity/Inhibition Fast Assay Kit
Used for measuring activity/inhibition of H3-K9 specific Histone Demethylases using cell/tissue extracts
Manufacturer:  Abnova™ KA0576
Catalog No. 89-027-139
Add to cart
Specifications
Specifications
Histone Demethylase (H3-K9 Specific) Activity/Inhibition Fast Assay Kit is use for measuring activity/inhibition of H3-K9 specific histone demethylases using cell/tissue extracts.
4mL HL1 (HDM Assay Buffer), 100μL HL2 (HDM Substrate), 50μL HL3 (HDM standard, 10 mM), 8mL HL4 (fluoro-developer), 8mL HL5 (fluoro-enhancer), 50μL HDM cofactor 1, 50μL HDM cofactor 2,
Histone Demethylase (H3-K9 Specific) activity/inhibition fast assay kit
Store HL2 at -20°C away from light. Store all other components at 4°C away from light. Tightly cap the HL4 after each use.
Functional Study
Histone Demethylase (H3-K9 Specific) Activity/Inhibition Fast Assay Kit
1 Kit