Learn More
Abnova™ HP (Rabbit) ELISA Kit
Enzyme-linked immunoassay for the quantitative determination of HP in Rabbit Serum or Plasma
Manufacturer:  Abnova™ KA1930
Catalog No. 89-028-645
Add to cart
Specifications
Specifications
HRP
HP (Rabbit) ELISA Kit is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of HP in rabbit serum or plasma.
Quantification
HP (Rabbit) ELISA Kit
1 Kit
Store the kit at 4°C.
Rabbit
Colorimetric
50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator (lyophilized)
HP (Rabbit) ELISA kit
Plasma, Serum
100μL