Learn More
Abnova™ HSPB1 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human HSPB1
Manufacturer:  Abnova™ KA1065
Catalog No. 89-027-203
Add to cart
Specifications
Specifications
62.5 to 4000pg/mL
Human
5pg/mL
Quantification
HSPB1 (Human) ELISA Kit
1 Kit
Store the kit at-20°C.
Biotin
Human HSPB1 ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of human HSPB1.
Colorimetric
96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Human HSPB1 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human HSPB1 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin-Biotin
HSPB1 (Human) ELISA kit
Body Fluid, Cell Culture Supernatant, Serum, Tissue Lysate
100μL