Learn More
Abnova™ Hydrogen Peroxide Assay Kit
To measure hydrogen peroxide in biological samples, functional study
Manufacturer:  Abnova™ KA0801
Catalog No. 89-028-095
Add to cart
Specifications
Specifications
Hydrogen Peroxide Assay Kit is used for measuring H2O2 in biological samples.
25mL Hydrogen Peroxide assay buffer, 0.2mL OxiRed™ probe (in anhydrous DMSO), 1 vial HRP, 0.1mL Hydrogen Peroxide standard (0.88M)
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
Hydrogen Peroxide Assay Kit