Abnova™ IFNG (Human), Biotin, ELISA Kit Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Abnova Corporation KA0135

 View more versions of this product

Catalog No. 89-100-096

  Check Availability
Add to Cart

    Specifications

    Product Type IFNG (Human) ELISA Kit
    Quantity 1 Kit
    Regulatory Status RUO
    Sample Type Cell culture supernatant, Plasma, serum
    Calibration Range 1.6 to 100pg/mL
    Conjugate Biotin
    Cross Reactivity Human

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit