Learn More
Abnova™ IGFBP1 (Human) ELISA Kit
In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human IGFBP1
Manufacturer:  Abnova™ KA1872
 View more versions of this product
Catalog No. 89-028-583
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Biotin
IGFBP1 (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IGFBP1.
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB one step Substrate reagent, 8mL stop solut
96 Assays
Cell Culture Supernatant, Plasma, Serum, Urine
96 Assays
Human
5pg/mL
Quantification
IGFBP1 (Human) ELISA Kit
For research use only (RUO)
Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
100μL