Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ IGFBP1 (Human) ELISA Kit
In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human IGFBP1
$1190.00
Specifications
Conjugate | Biotin |
---|---|
Cross Reactivity | Human |
Detection Method | Colorimetric |
Quantity | 96 Assays |
Regulatory Status | For research use only (RUO) |
Catalog Number | Mfr. No. | Detection Limit | Includes | Sample Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detection Limit | Includes | Sample Type | Price | Quantity & Availability | |||
89-100-760
|
Abnova™
KA3053 |
1pg/mL | One 96-well plate pre-coated with anti-Human IGFBP-1 antibody, Lyophilized IGFBP-1 standards (10ng x 2 ), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human IGFBP-1 antibody (Concentrated), Dilution: 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130ul ), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL) | Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate |
Each for $1,190.00
|
|
|||
89-028-583
|
Abnova™
KA1872 |
5pg/mL | 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB one step Substrate reagent, 8mL stop solut | Cell Culture Supernatant, Plasma, Serum, Urine | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Specifications
Biotin | |
Colorimetric | |
For research use only (RUO) | |
Quantification | |
96 |
Human | |
96 Assays | |
100 μL | |
IGFBP1 (Human) ELISA Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title