Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgG1 (Mouse) ELISA Kit

Colorimetric immunometric Enzyme Immunoassay kit for the quantitative measurement of Mouse IgG1 in culture supernatants and Serum of Mouse origin

Manufacturer:  Abnova™ KA1011

 View more versions of this product

Catalog No. 89-028-179


Add to cart

Specifications

Specifications

7.8 to 250ng/mL
Mouse
1.79ng/mL
Quantification
IgG1 (Mouse) ELISA Kit
1 Kit
Serum, Tissue Culture Medium
96 Assays
HRP
IgG1 (Mouse) ELISA Kit is a colorimetric immunometric enzyme immunoassay kit for the quantitative measurement of mouse IgG1 in culture supernatants and serum of mouse origin.
Colorimetric
96-well microwell, 6mL IgG1 Conjugate, 50mL Assay Buffer concentrate, 100mL Wash Buffer concentrate, 0.25mL standard, 12mL TMB Substrate, 11mL stop solution
IgG1 (Mouse) ELISA kit
RUO
Store the kit at 4°C.
50μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit