Learn More
Abnova™ IgG2a (Mouse) ELISA Kit
Colorimetric immunometric Enzyme Immunoassay for the quantitative measurement of IgG2a in plasma and serum
Manufacturer:  Abnova™ KA1013
 View more versions of this product
Catalog No. 89-028-177
Add to cart
Specifications
Specifications
7.18 to 250ng/mL
Mouse
0.32ng/mL
Quantification
IgG2a (Mouse) ELISA Kit
1 Kit
Serum, Tissue Culture Medium
96 Assays
HRP
IgG2a (Mouse) ELISA Kit is a colorimetric immunometric enzyme immunoassay kit for the quantitative measurement of IgG2a in culture supernatants and serum of mouse origin.
Colorimetric
96-well microwell, 5mL IgG2a Conjugate, 50mL Assay Buffer concentrate, 100mL Wash Buffer concentrate, 1 standard vial, 10mL TMB Substrate, 10mL stop solution
IgG2a (Mouse) ELISA kit
RUO
Store standard at -20°C in the original bottle. Store all other components at 4°C. Avoid repeated freezing and thawing.
50μL