Learn More
Abnova™ IL17AF ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of free IL17AF in the supernatant of activated cells
Manufacturer:  Abnova™ KA1397
Catalog No. 89-024-517
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
IL17AF ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of free IL17AF in the supernatant of activated cells.
Quantification
IL17AF ELISA Kit
1 Kit
Store the standard at -20°C. Store all other components at 4°C.
Human
Colorimetric
250μL 100X Capture Antibody, 250μL 100X detection Antibody, 4 x 0.5g BSA, 50mL Wash Buffer 20X, 25mL Coating Buffer, 20mL 10X Blocking Buffer, 10μL HRP Conjugated Streptavidin, 2 x 12mL ABTS Substrate,
IL-17AF ELISA kit
Cell Culture Supernatant
100μL