Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ IL18 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human IL-18
$1307.00
Specifications
Calibration Range | 15.6 to 1000pg/mL |
---|---|
Conjugate | Biotin |
Cross Reactivity | Human |
Detection Limit | 1pg/mL |
Detection Method | Colorimetric |
Catalog Number | Mfr. No. | Includes | Sample Type | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Sample Type | Content And Storage | Price | Quantity & Availability | |||
89-027-125
|
Abnova™
KA0561 |
96-well Antibody coated microwell, 2 vials x 10ng recombinant Human IL18 standard (lyophilized), 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human IL18 Antibody, 12mL Antibody Diluent Buffer, 130uL Advin-Biotin Peroxidase Complex,12mL ABC diluent buffer, 10mL TMB color developing agent, 10mL TMB stop solution | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum | Store the kit at -20°C |
Each for $1,307.00
|
|
|||
89-100-782
|
Abnova™
KA3075 |
96-well plate pre-coated with anti-Human IL-18 antibody, Lyophilized IL-18 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human IL-18 antibody (Concentrated Dilution: 1:100) (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100) (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL) | Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate | Store at 4°C for 4 months, or at -20°C for 8 months. |
Each for $1,307.00
|
|
|||
Specifications
15.6 to 1000pg/mL | |
Human | |
Colorimetric | |
100 μL | |
IL18 (Human) ELISA Kit |
Biotin | |
1pg/mL | |
For research use only (RUO) | |
Quantification | |
96 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title