Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL18R1 (Human) ELISA Kit

Sandwich enzyme immunoassay for the quantitative measurement of human IL18R1 in cell culture supernates, plasma (EDTA, heparin) and serum samples

Manufacturer:  Abnova™ KA4297

Catalog No. 89-139-163


Add to cart

Description

Description

  • Sample Type: Cell culture supernates, serum, plasma (heparin, EDTA)
  • Sample Volume: 100μL
  • Conjugate: Biotin
  • Reactivity: Human
  • Detection Method: Colorimetric
  • Calibration Range: 31.2 to 2000 pg/mL
  • Limit of Detection: <10 pg/mL

  • Applications: Quantification

  • Storage: Store the kit at -20°C.

Specifications

Specifications

Sandwich ELISA
Biotin
ELISA Kit
31.2 to 2000 pg/mL
Quantification
For research use only (RUO)
Store the kit at -20°C
Human
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
96-well Plate
No detectable cross-reactivity with other relevant proteins
Natural and recombinant human IL-18R1
96 Assays
IL18R1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL18R1.
8809
100 μL
IL18R1
1 Kit
<10 pg/mL
Colorimetric
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only (RUO).