Abnova™ IL6 (Human) ELISA Kit Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Abnova Corporation KA0123

 View more versions of this product

Catalog No. 89-022-604

  Check Availability
Add to Cart

    Specifications

    Cross Reactivity Human
    Detection Limit 0.92pg/mL
    Detection Method Colorimetric
    For Use With (Application) Quantitative measurement of Human IL-6, quantification
    Includes 96-well Antibody coated microwell, 100μL Biotin Conjugate Antibody, 150μL Streptavidin HRP, 2 vials standard (lyophilized), 12mL sample diluent, 5mL Assay Buffer concentrate 20X, 50mL Wash Buffer 20X concentrate, 15mL S
    Product Type IL-6 (Human) ELISA kit
    Quantity 96 Assays
    Sample Type Cell Culture Supernatant, Plasma (Citrate, EDTA, Heparin), Serum
    Storage Requirements 4°C
    Sufficient For 96 Assays
    Volume (Metric) Sample 50μL
    Calibration Range 1.56 to 100pg/mL
    Conjugate Biotin

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit