Learn More
Abnova™ In Situ BrdU DNA Fragmentation Assay Kit
In Situ BrdU DNA Fragmentation Assay Kit using Br-dUTP for detecting DNA fragmentation by fluorescein.
Manufacturer:  Abnova™ KA0773
Catalog No. 89-028-007
Add to cart
Description
Description
  • Internucleosomal DNA fragmentation is a hallmark of apoptosis in mammalian cells
  • Provides complete components including positive and negative control cells for conveniently detecting DNA fragmentation by fluorescence microscopy or flow cytometry
  • The kit utilizes Br-dUTP (bromolated deoxyuridine triphosphate nucleotides) which is more readily incorporated into DNA strand breaks than other larger ligands (e.g., fluorescein, biotin or digoxigenin)
  • The greater incorporation gives rise to brighter signal when the Br-dUTP sites are identified by a fluorescein labeled anti-BrdU monoclonal antibody
  • The assay is suitable for studying apoptosis with GFP transfected cells
  • For research use only
Specifications
Specifications
In Situ BrdU DNA Fragmentation Assay Kit using Br-dUTP for detecting DNA fragmentation by fluorescein.
Liquid (Ethanol), Solid (Sodium Phosphate), Powder (Sodium Azide), Powder (Cobalt Chloride), Solid (Cacodylic Acid), Powder (Fluorescein), Powder (Propidium iodide)
Store the kit at -20°C.
1 Kit
Functional Study