Learn More
Abnova™ In Situ BrdU DNA Fragmentation Assay Kit (IHC)
In Situ BrdU DNA Fragmentation Assay Kit (IHC) using Br-dUTP for detecting DNA fragmentation by IHC.
Manufacturer:  Abnova™ KA0775
Catalog No. 89-028-005
Add to cart
Description
Description
  • A two-color TUNEL (Terminal deoxynucleotide transferase dUTP Nick End Labeling) assay for labeling DNA breaks to detect apoptotic cells by immunohistochemistry
  • Kit contains positive/negative control slides for assessing reagent performance; reaction and blocking buffers for processing individual steps in the assay; proteinase K; terminal deoxynucleotidyl tranferase enxyme, etc
  • After initial defrosting, the 5x Reaction Buffer and Anti-BrdU-Biotin mAb should be stored at 4°C and Methyl Green should be stored at room temperature
  • For research use only
Specifications
Specifications
In Situ BrdU DNA Fragmentation Assay Kit (IHC) using Br-dUTP for detecting DNA fragmentation by IHC.
Liquid (H2O2), Powder (DAB), Liquid (Glycerol), Solid (Cacodylic Acid), Powder (Cobalt Chloride), Crystalline (Sodium Azide)
Functional Study, Immunohistochemistry
1 Kit
Horseradish Peroxidase Streptavidin
Store the kit at -20°C.