Learn More
Abnova™ In Situ DNA Fragmentation Assay Kit
In Situ DNA Fragmentation Assay Kit is a TUNEL (Terminal deoxynucleotide transferase dUTP Nick End Labeling) assay for detecting DNA fragmentation.
Manufacturer:  Abnova™ KA0774
Catalog No. 89-028-006
Add to cart
Description
Description
  • Internucleosomal DNA fragmentation is a hallmark of apoptosis in mammalian cells
  • Provides complete components including positive and negative control cells for conveniently detecting DNA fragmentation by fluorescence microscopy or flow cytometry
  • The TUNEL-based detection kit utilizes terminal deoxynucleotidyl transferase (TdT) to catalyze incorporation of fluorescein-12-dUTP at the free 3’-hydroxyl ends of the fragmented DNA
  • The fluorescein-labeled DNA can then be observed by fluorescence microscopy or analyzed by flow cytometry
  • Shelf life is 1 year from the date of the product shipment, under proper storage conditions
  • For research use only
Specifications
Specifications
In Situ DNA Fragmentation Assay Kit is a TUNEL (Terminal deoxynucleotide transferase dUTP Nick End Labeling) assay for detecting DNA fragmentation.
Liquid (Ethanol), Solid (Sodium Phosphate), Powder (Sodium Azide), Powder (Cobalt Chloride), Solid (Cacodylic Acid), Powder (FITC), Powder (Propidium iodide)
Store the kit at -20°C.
1 Kit
Functional Study