Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Insulin (Human) ELISA Kit
For quantitative measurement Insulin in Human Serum or Plasma
$791.00
Specifications
Cross Reactivity | Human |
---|---|
Detection Method | Colorimetric |
Product Type | Insulin (Human) ELISA kit |
Includes:
- Microwells coated with Insulin MAb 12x8x1
- Insulin Standard 1:1 vial (Ready to use) 2mL
- Insulin Standards 2-6: 5 vials (Ready to use) 1mL
- Insulin Enzyme Conjugate: 1 vial 1mL
- Assay Diluent: 1 bottle (Ready to use) 14mL
- TMB Substrate: 1 bottle (Ready to use) 12mL
- Stop Solution: 1 bottle (Ready to use) 12mL
- 20X Wash concentrate: 1 bottle 25mL
Catalog Number | Mfr. No. | Calibration Range | For Use With (Application) | Sample Type | Content And Storage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Calibration Range | For Use With (Application) | Sample Type | Content And Storage | Price | Quantity & Availability | ||
89-101-131
|
Abnova™
KA3810 |
0 to 140μIU/mL | Quantification | Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysates | Store the kit at 4°C |
Each for $791.00
|
|
||
89-024-462
|
Abnova™
KA0921 |
6.25 to 100μIU/mL | Quantitative measurement Insulin in Human Serum or Plasma, quantification | Plasma, Serum | 4°C | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||
Specifications
Human | |
Colorimetric | |
Insulin (Human) ELISA kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title