Learn More
Abnova™ Isocitrate Assay Kit
Used for quantifying isocitrate and functional study
Manufacturer:  Abnova™ KA0865
Catalog No. 89-024-420
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
1 to 20nm
Functional Study
1 Kit
Store the kit at -20°C.
Isocitrate Assay Kit
Isocitrate Assay Kit is used for quantifying isocitrate.
25mL Isocitrate assay buffer, 0.2mL Isocitrate enzyme mix, 1 vial substrate mix, 1 vial Isocitrate standard
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric