Learn More
Abnova™ JMJD2 Demethylase Activity/Inhibition Assay Kit (Colorimetric)
To screen JMJD2 demethylase inhibitors, functional study
Manufacturer:  Abnova™ KA1526
Catalog No. 89-028-221
Add to cart
Specifications
Specifications
JMJD2 Demethylase Activity/Inhibition Assay Kit (Colorimetric) is use for screening JMJD2 demethylase inhibitors.
14mL JA1 (wash buffer 10X), 4mL JA2 (JMJD2 assay buffer), 60μL JA3 (JMJD2 substrate, 50μg/mL), 10μL JA4 (JMJD2 assay standard, 50μg/mL), 5μL JA5 (Capture antibody, 1
Cell lysate
Colorimetric
Functional Study
1 Kit
Store strips at 4°C. Store JA3, JA4 and JA6 at -20°C. Store remaining components at room temperature away from light.
JMJD2 Demethylase Activity/inhibition Assay Kit