Learn More
Abnova™ JNK Activity Assay Kit
JNK Activity Assay Kit is used for detecting JNK.
Manufacturer:  Abnova™ KA0889
Catalog No. 89-024-444
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • c-Jun N-terminal kinase (JNK) is one of the several main MAP kinase groups identified in mammals
  • Recent evidences suggest that activation of JNK may play an important role in neuronal apoptosis and other physiological and pathological processes
  • The JNK Activity Assay Kit utilizes a JNK-specific antibody to immunoprecipitate JNK from cell lysate
  • Phosphorylation of the c-Jun can be analyzed by Western blot analysis using a phospho-c-Jun specific antibody
  • The kit specifically detects JNK, other kinase activity would not be detected
  • Store kit at -20°C
  • For research use only
Specifications
Specifications
1 Kit
JNK Activity Assay Kit is used for detecting JNK.
Store the kit at -20°C.
Functional Study, Immunoprecipitation, Western Blot