Learn More
Abnova™ JNK Activity Screening Kit
JNK Activity Screening Kit is used for detecting JNK activity.
Manufacturer:  Abnova™ KA0888
Catalog No. 89-024-443
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • c-Jun N-terminal kinase (JNK) is one of the several main MAP kinase groups identified in mammals
  • Recent evidences suggest that activation of JNK may play an important role in neuronal apoptosis and other physiological and pathological processes
  • The JNK Activity screening kit utilizes an N-terminal c-Jun (1-79) fusion protein bound to glutathion sepharose beads to selectively “pull down” JNK from cell lysates
  • c-Jun phosphorylation is then measured by Western blot analysis using a phospho-c-Jun specific antibody
  • For research use only
Specifications
Specifications
1 Kit
JNK Activity Screening Kit is used for detecting JNK activity.
Store the kit at -20°C.
Functional Study, Immunoprecipitation, Western Blot