Learn More
Abnova™ KNG1 (Human) ELISA Kit
Competitive Enzyme Immunoassay for the quantitative measurement of Human KNG1 in Plasma and Serum
Manufacturer:  Abnova™ KA1040
Catalog No. 89-028-152
Add to cart
Specifications
Specifications
0.125 to 4μg/mL
Human
0.1μg/mL
Quantification
KNG1 (Human) ELISA Kit
1 Kit
Store components of the kit at 4°C or -20°C as described in the protocol.
25μL
HRP
KNG1 (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human KNG1 in plasma and serum.
Colorimetric
96-well Antibody coated microwell, 2μg Human KNG2 standard, 1 vial 50X Biotinylated KNG1, 30mL 10X MIX diluent concentrate, 30mL Wash Buffer 20X concentrate, 80μL Streptavidin-Peroxidase Conjugate (SP Conjugate),, 8mL c
KNG1 (Human) ELISA kit
Plasma, Serum
96 Assays