Abnova™ L-Alanine Assay Kit Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Abnova Corporation KA0863

 View more versions of this product

Catalog No. 89-024-418

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Calibration Range 24 to 76μg/mL
    For Use With (Application) Used for detecting alanine and functional study
    Includes 25mL Alanine assay buffer, 0.2mL Alanine probe (in DMSO), 1 vial Alanine Converting enzyme (lyophilized), 1 vial Alanine development mix (lyophilized), 1 vial Alanine standard (10μM, lyophilized)
    Quantity 100 Assays
    Sample Type Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
    Storage Requirements -20°C
    Detection Method Colorimetric, Fluorometric
    Type L-Alanine Assay Kit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit