Learn More
Abnova™ L-Amino Acid Quantitation Kit
Used for detecting L-amino acid
Manufacturer:  Abnova™ KA0859
Catalog No. 89-024-414
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
L-Amino Acid Quantitation Kit is used for detecting L-amino acid.
25mL L-Amino Acid assay buffer, 1 vial L-Amino Acid probe, 400μL DMSO (anhydrous), 1 vial L-Amino Acid enzyme mix, 300μL L-Amino Acid standard
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
L-Amino Acid Quantitation Kit