Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ L-Carnitine Assay Kit
Used for measuring free L-Carnitine and functional study
Supplier: Abnova™ KA0860
Specifications
Store the kit at -20°C. | |
Colorimetric, Fluorometric | |
25mL Carnitine assay buffer, 0.2mL Carnitine probe (in DMSO), Carnitine Converting enzyme (lyophilized), 400μL Carnitine substrate mix, Carnitine development mix (lyophilized), Carnitine standard (10μM, lyophilized) | |
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
L-Carnitine Assay Kit is used for measuring free L-carnitine. | |
Functional Study | |
1 Kit | |
Detection Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction