Learn More
Abnova™ Lactate Assay Kit II
Detects L(+)-Lactate in biological samples such as serum or plasma, cells, culture and fermentation media at a level of 0.02 to 10 mM. There is no need for pretreatment or purification of samples.
Manufacturer:  Abnova™ KA0834
Catalog No. 89-028-066
Add to cart
Specifications
Specifications
0.002 to 10mm
Functional Study
1 Kit
Store the kit at -20°C.
Lactate Assay Kit II
Lactate Assay Kit II is used for detecting L(+)-Lactate.
25mL Lactate assay buffer, Lactate enzyme mix (lyophilized), Lactate substrate mix (lyophilized), 100μL L(+) Lactate standard (100mM)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric