Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lactate Dehydrogenase Assay Kit

Quantifies LDH activity in variety of biological samples such as serum or plasma, cells, culture medium and fermentation, etc. The assay is quick, convenient, and sensitive. The kit can detect 1 to 100 mU/mL of LDH directly in samples.

Manufacturer:  Abnova™ KA0878

Catalog No. 89-024-433


Due to product restrictions, please to purchase or view availability for this product.


Includes:
  • LDH Assay Buffer 50mL
  • LDH Substrate Mix (lyophilized) 1 vial
  • NADH Standard (0.5µmol; lyophilized) 1 vial
  • LDH Positive Control 20µL

Description

Description

Lactate Dehydrogenase Assay Kit is used for quantifying LDH activity

Functional Study

Specifications

Specifications

1 Kit
Store the kit at -20°C.
1 to 100mU/mL
Lactate Dehydrogenase Assay Kit is used for quantifying LDH activity.
Colorimetric
Functional Study
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit