Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lactoferrin Ab ELISA Kit

Indirect solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Lactoferrin in human serum or plasma

Manufacturer:  Abnova™ KA1267

Catalog No. 89-024-559


Add to cart

Description

Description

  • Sample Type: Plasma, Serum
  • Sample Volume: 100µL

Applications:

Quantification

Specifications

Specifications

HRP
Anti-Lactoferrin Ab ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Lactoferrin in human serum or plasma.
Colorimetric
6 Vials, 1.5mL combined calibrator with IgG class Anti-lactoferrin, 2 vials x 1.5mL Anti-lactoferrin controls, 1 vial x 20mL Sample Buffer, 1 vial x 15mL Enzyme Conjugate solution, 1 vial x 15mL TMB Substrate solution, 1 vial x 15mL stop solution,, one 96-well plate, 1 vial x 20mL wash solution
Lactoferrin Ab ELISA kit
Plasma, Serum
96 Assays
Human
0.5U/mL
Quantification
Lactoferrin Ab ELISA Kit
1 Kit
Store the kit at 4°C.
100μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit