Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LDH-Cytotoxicity Assay Kit II

To utilize the advanced WST reagent for a fast and more sensitive detection of LDH released from damaged cells, functional study

Manufacturer:  Abnova™ KA0786

Catalog No. 89-028-107


Add to cart

Includes:
  • WST substrate mix, 1 vial
  • LDH assay buffer, 50mL
  • Cell lysis solution, 6mL
  • Stop solution, 6mL
  • LDH, 20µL

Specifications

Specifications

LDH-Cytotoxicity Assay Kit II is used for utilizing the advanced WST reagent for a fast and more sensitive detection of LDH released from damaged cells.
1 vial WST substrate mix, 50mL LDH assay buffer, 6mL cell lysis solution, 6mL stop solution, 20μL LDH
Cell
Colorimetric
Functional Study
1 Kit
Store the kit at -20°C.
LDH-Cytotoxicity Assay Kit II
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit