Learn More
Abnova™ LDH-Cytotoxicity Assay Kit
To quantify cytotoxicity based on the measurement of activity of lactate dehydrogenase (LDH) released from damaged cells, functional study
Manufacturer:  Abnova™ KA0785
Catalog No. 89-028-108
Add to cart
Specifications
Specifications
LDH-Cytotoxicity Assay Kit is used for quantitating cytotoxicity based on the measurement of activity of lactate dehydrogenase (LDH) released from damaged cells.
1 vial catalyst (lyophilized), 45mL dye solution
Cell
Colorimetric
Functional Study
1 Kit
Store the kit at -20°C.
LDH-Cytotoxicity Assay Kit