Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Lipase Assay Kit
Used for measuring lipase activity and functional study
Supplier: Abnova™ KA0877
Includes:
- Lipases assay buffer, 25mL
- OxiRed probe (in DMSO), 0.2mL
- Enzyme mix (lyophilized), 1 vial
- Lipase substrate, 0.4mL
- Glycerol standard (100mM), 0.2mL
- Lipase positive control (lyophilized), 1 vial
Specifications
Store the kit at -20°C. | |
Colorimetric | |
25mL Lipases assay buffer, 0.2mL OxiRed™ (in DMSO), 1 vial enzyme mix (lyophilized), 0.4mL Lipase substrate, 0.2mL Glycerol standard (100mM), 1 vial Lipase positive control (lyophilized) | |
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine, Food Sample, Growth Medium |
Lipase Assay Kit is used for measuring lipase activity. | |
Functional Study | |
1 Kit | |
Detection Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction