Learn More
Abnova™ Live-Dead Cell Staining Kit
Provides the ready-to-use reagents for convenient discrimination between live and dead cells.
Supplier:  Abnova™ KA0901
Catalog No. 89-024-450
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
The kit uses Live-Dye™, a cell-permeable green fluorescent dye, to stain live cells. Dead cells can be easily stained by propidium iodide (PI), a cell non-permeable red fluorescent dye.
Specifications
Specifications
Store the kit at -20°C.
Functional Study
1 Kit
Live-Dead Cell Staining Kit is used for discriminating live and dead cells.
50μL Solution A (1mM Live-dye), 50μL solution B (2.5mg/mL PI), 50mL Staining buffer
Cell