Learn More
Abnova™ LKM-1 Ab ELISA Kit
Detection of antibodies against Liver-Kidney Microsomes in Human Serum, qualitative
Manufacturer:  Abnova™ KA1280
Catalog No. 89-024-572
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
LKM-1 Ab ELISA Kit is a solid phase enzyme immunoassay employing human recombinant cytochrome p450 IID6 for the detection of antibodies against liver-kidney microsomes (LKM) in human serum.
Colorimetric
1 Vial x 20mL 5X Sample Buffer, 50X Wash Buffer, 1 vial x 1.5mL negative control, 1 vial x 1.5mL positive control, 1 vial x 1.5mL cut-off calibrator, one 96-well plate, 6 vials x 1.5mL calibrator, 1 vial x 15mL Conjugate, 1 vial x 15mL TMB Substrate,, 1 vial x 15mL stop solution
LKM-1 Ab ELISA kit
Serum
96 Assays
Human
1U/mL
Qualitative
LKM-1 Ab ELISA Kit
1 Kit
Store the kit at 4°C.
100μL