Learn More
Abnova™ LSD1 Demethylase Activity/Inhibition Assay Kit (Colorimetric)
To screen LSD1 demethylase inhibitors, functional study
Manufacturer:  Abnova™ KA1525
Catalog No. 89-028-220
Add to cart
Specifications
Specifications
LSD1 Demethylase Activity/Inhibition Assay Kit (Colorimetric) is use for screening LSD1 demethylase inhibitors.
14mL LD1 (wash buffer 10X), 4mL LD2 (LSD1 assay buffer), 60μL LD3 (LSD1 substrate, 50μg/mL), 10μL LD4 (LSD1 assay standard, 50μg/mL), 5μL LD5 (Capture antibody, 1000
Cell lysate
Colorimetric
Functional Study
1 Kit
Store LD1, LD5, LD7, LD8 and 8-Well Assay Strips at 4°C away from light. Store LD3, LD4, and LD6 at -20°C away from light. Store remaining components (LD2, LD9, and Adhesive Covering Firm) at room temperature away from light.
LSD1 Demethylase Activity/inhibition Assay Kit